Learn More
Selleck Chemical LLC Rasagiline Mesylate S2102-50mg
SDP
Supplier:  Selleck Chemical LLC S210250MG
Encompass_Preferred
Rasagiline Mesylate (TVP-1012 (R)-AGN1135 mesylate TVP1012 mesylate) is a new MAO-B inhibitor for the treatment of idiopathic Parkinson s disease
Catalog No. 50-311-1130