Learn More
Medchemexpress LLC Rasarfin | 674359-73-0 | 99.6% | 425.91 | 1 MG
SDP
Supplier:  Medchemexpress LLC HY1399501MG
Encompass_Preferred
Rasarfin is a dual Ras and ARF6 inhibitor for research use only. It potently inhibits agonist-induced ERK1/2 signaling by GPCRs, and MAPK and Akt signaling by EGFR. This compound prevents cancer cell proliferation and blocks GPCR internalization through inhibition of ARF6. Treatment with Rasarfin results in a dose-dependent reduction in MDA-MB-231 cells.
Catalog No. 50-003-51228