Learn More
Medchemexpress LLC Rasarfin | 674359-73-0 | 99.6% | 425.91 | 1 ML
SDP
Supplier:  Medchemexpress LLC HY13995010MM/1ML
Encompass_Preferred
Rasarfin is a dual Ras and ARF6 inhibitor. It potently inhibits agonist-induced ERK1/2 signaling by GPCRs, and MAPK and Akt signaling by EGFR, preventing cancer cell proliferation. It blocks GPCR internalization by inhibiting ARF6 and leads to a dose-dependent reduction in MDA-MB-231 cells. Rasarfin inhibits the recruitment of the clathrin adaptor AP-2 to β-arrestin2 and receptor internalization. This product is for research use only.
  • Inhibits agonist-induced ERK1/2 signaling by GPCRs
  • Inhibits MAPK and Akt signaling by EGFR
  • Prevents cancer cell proliferation
  • Blocks GPCR internalization through inhibition of ARF6
  • Reduces MDA-MB-231 cells in a dose-dependent manner
  • Inhibits recruitment of clathrin adaptor AP-2 to β-arrestin2 and receptor internalization
Catalog No. 50-003-51144