Learn More
Medchemexpress LLC Rasarfin | 674359-73-0 | 99.6% | 425.91 | 50 MG
SDP
Supplier:  Medchemexpress LLC HY13995050MG
Encompass_Preferred
Rasarfin is a dual inhibitor of Ras and ARF6. It potently inhibits agonist-induced ERK1/2 signaling by GPCRs, and MAPK and Akt signaling by EGFR. It also prevents cancer cell proliferation and blocks GPCR internalization through inhibition of ARF6. Treatment with Rasarfin leads to a dose-dependent reduction in MDA-MB-231 cells.
  • Potently inhibits agonist-induced ERK1/2 signaling by GPCRs
  • Inhibits MAPK and Akt signaling by EGFR
  • Prevents cancer cell proliferation
  • Blocks GPCR internalization through inhibition of ARF6
  • Inhibits recruitment of the clathrin adaptor AP-2 to β-arrestin2 and receptor internalization
Catalog No. 50-003-51067