Learn More
Aviva Systems Biology Corp Rasl10a Antibody - N-terminal region (ARP52194_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP52194_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against Rasl10a. It was validated on Western Blot. . Alternate names: 2210403B10Rik, AI852688, MGC130093, MGC130094ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-285-658