Learn More
Med Vet International Miltex Fomon Rasp, 8.25in., Double Ended
Supplier:  Med Vet International MIL21369
Encompass_Preferred
The Miltex 21-369 Fomon Rasp is double ended with coarse teeth with backwards cutting. It is made of German surgical grade stainless steel. The Fomon Rasp is a multipurpose veterinary surgical instrument for bone contouring in nasal reconstructive procedures.
Catalog No. 50-283-0078