Learn More
Medchemexpress LLC HY-N1426 100mg Medchemexpress, Raspberry ketone CAS:5471-51-2 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYN1426100MG
Encompass_Preferred
Medchemexpress, HY-N1426 100mg Raspberry ketone CAS:5471-51-2 Raspberry ketone is a major aromatic compound of red raspberry, widely used as a fragrance in cosmetics and as a flavoring agent in foodstuff; also shows PPAR-α agonistic activity. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-3829