Learn More
American Research Products Inc Rat alcohol dehydrogenase ADH
Supplier:  American Research Products Inc CSBE11180R
Encompass
Rat alcohol dehydrogenase ADH ELISA Kit from CUSABIO Detection Range 0 312 ng/mL-20 ng/mL Sensitivity 0 078 ng/mL Sample serum plasma tissue homogenates Method Sandwich
Catalog No. 50-003-26136