Learn More
American Research Products Inc Rat Anti Cyclic Citrullinated
Supplier:  American Research Products Inc CSBE13830R
Encompass
Rat Anti-Cyclic Citrullinated Peptide Antibody (Anti-CCP-antibody) ELISA Kit from CUSABIO Detection Range 3 12 IU/mL--200 IU/mL Sensitivity 0 78 IU/mL Sample serum plasma Method Sandwich
Catalog No. 50-003-29362