Learn More
American Research Products Inc Rat anti Mullerian hormone AM
Supplier:  American Research Products Inc CSBE11162R
Encompass
Rat anti-Mullerian hormone (AMH) ELISA kit from CUSABIO Detection Range 0 2 ng/mL-15 ng/mL Sensitivity 0 1 ng/mL Sample serum plasma tissue homogenates Method Competitive
Catalog No. 50-003-26205