Learn More
American Research Products Inc Rat Antithrombin III AT III
Supplier:  American Research Products Inc CSBE13885R
Encompass
Rat Antithrombin III (AT III) ELISA Kit from CUSABIO Detection Range 39 ng/mL-2500 ng/mL Sensitivity 9 75 ng/mL Sample serum plasma tissue homogenates Method Sandwich
Catalog No. 50-003-29401