Learn More
Sino Biological Recombinant Anti-BCAM Antibody, Rabbit Monoclonal
Supplier:  Sino Biological 80222R113100
Encompass_Preferred
This antibody was obtained from a rabbit immunized with purified, recombinant Rat BCAM / CD239 (Catalog#80222-R08H; Q9ESS6; Met1-Ala543).
Catalog No. 50-255-2809