Learn More
Lonza Walkersville Rat Brain Hypothalamus Neurons
Supplier:  Lonza Walkersville RHTH507
Encompass
The hypothalamus is responsible for many metabolic processes and other activities of the autonomic nervous system. It synthesizes and secretes neurohormones, that stimulate or inhibit the secretion of pituitary hormones. The hypothalamus controls body temperature, hunger, thirst, fatigue, and circadian cycles. Rat hypothalamus neurons are a useful tool in the study of neuroendocrine-related diseases and disorders.Benefits:Ready-To-Use No tissue dissection or animal handling required Increased data reproducibilityCharacteristic morphology and staining tests confirm tissue origin and cell typeNegative for mycoplasma, bacteria, yeast and fungiCertificate of Analysis (CoA) is available for every batchApplications:NeurogenesisNeurotransmitter FunctionGene ExpressionSignaling PathwaysElectrophysiologyNeurotoxicityDrug/Compound ScreeningAdvanced Cell Culture ModelsContent:≥ 2,000,000 cryopreserved cells per ampule
Catalog No. NC2381522