Learn More
Sino Biological Anti-BTLA Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 104829T32100
Encompass_Preferred
Produced in rabbits immunized with a synthetic peptide corresponding to the center region of the Rat BTLA/CD272, and purified by antigen affinity chromatography.
Catalog No. 50-241-0580