Learn More
American Research Products Inc Rat C ReactiVe Protein CRP EL
Supplier:  American Research Products Inc CSBE07922R
Encompass
Rat C-ReactiVe Protein CRP ELISA Kit from CUSABIO Detection Range 31 25 ng/mL-2000 ng/mL Sensitivity 7 81 ng/mL Sample serum plasma cell culture supernates tissue homogenates cell lysates Method Sandwich
Catalog No. 50-003-26094