Learn More
American Research Products Inc Rat C X C motif chemokine 3 C
Supplier:  American Research Products Inc CSBEL006249RA
Encompass
Rat C-X-C motif chemokine 3 (CXCL3) ELISA kit from CUSABIO Detection Range 15 6 pg/ml - 1000 pg/ml Sensitivity 3 9 pg/ml Sample serum plasma tissue homogenates Method Sandwich
Catalog No. 50-003-28143