Learn More
American Research Products Inc Rat Carcinoembryonic Antign C
Supplier:  American Research Products Inc CSBE13926R
Encompass
Rat Carcinoembryonic Antign CEA ELISA Kit from CUSABIO Detection Range 0 312 ng/mL-20 ng/mL Sensitivity 0 078 ng/mL Sample serum plasma tissue homogenates cell lysates Method Sandwich
Catalog No. 50-003-27475