Learn More
Lonza Walkersville Rat Cardiomyocyte Growth Medium (rCGM)BulletKit
Supplier:  Lonza Walkersville CC4515
Encompass
RCGM Rat Cardiomyocyte Growth Medium is specifically formulated for the growth and survival of rat neonatal cardiomyocytes in culture.Lonza offers customers a fully validated and optimized cardiomyocyte medium and growth factors to complement our Rat Cardiomyocytes, R-CM-561, as a complete system to study cardiac biology/toxicology.Content:1 x CC-3275 RCBM Basal Medium, 200 mL1 x CC-4516 SingleQuots Kit (growth factors and supplements)
Catalog No. NC9756315