Learn More
Raybiotech Inc Rat CD80 (B7-1) ELISA Kit
Supplier:  Raybiotech Inc ELRCD801
Encompass_Preferred
For serum, plasma, and cell culture supernatant samples. Sensitivity: 1.23 ng/ml. Pre-coated 96-well strip plate with additional reagents to allow for use in multiple experiments. Specificity: Cross Reactivity: This ELISA kit shows no cross-reactivity with the following cytokines tested: rat Activin A, Adiponectin, SLAMF2, CTACK, Decorin, Eotaxin-1, EphA5, EPO, FGF-BP, Flt-3L, Galectin-1, Galectin-3, Gas-1, GFRalpha-1, gp-130, HGF, IL-1R6, IL-1ra, IL-2Ralpha, IL-3, IL-7, IL-17F, IL-22, JAM-A, MIP-1alpha, Neuropilin-1, Neuropilin-2, Nope, Notch-1, Notch-2, P-Cadherin, Prolactin, RANTES, SCF, TIM-1 (KIM-1), TIMP-2, TREM-1, TWEAK R.
Catalog No. 50-195-0223