Learn More
American Research Products Inc Rat Ciliary Neurotrophic Facto
Supplier:  American Research Products Inc CSBE04528R
Encompass
Rat Ciliary Neurotrophic Factor CNTF ELISA Kit from CUSABIO Detection Range 15 6 pg/mL-1000 pg/mL Sensitivity 3 9 pg/mL Sample serum plasma tissue homogenates Method Sandwich
Catalog No. 50-003-26300