Learn More
ACROBiosystems ACROBiosystems Rat CLEC12A / MICL / CLL-1 Protein, His Tag
Supplier:  ACROBiosystems CLAR5248100UG
Encompass_Preferred
Recombinant Protein;100UG;Rat CLEC12A, His Tag (CLA-R5248) is expressed from human 293 cells (HEK293). It contains AA Tyr 65 - Leu 269 (Accession # B4F798-1).;This protein carries a polyhistidine tag at the N-terminus. The protein has a calculated MW of 26.0 kDa. The protein migrates as 32-45 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Catalog No. 50-210-7384