Learn More
ACROBiosystems Rat Complement C2 25ug
Supplier:  ACROBiosystems CO2R82E625UG
Encompass_Preferred
Biotinylated Rat Complement C2 Protein HisAvitag (CO2-R82E6) is expressed from human 293 cells (HEK293). It contains AA Ala 19 - Leu 758 (Accession # Q6MG73).
Catalog No. 50-253-5224