Learn More
Alpco Ltd Rat Complement Factor 3 ELISA
SDP
Supplier:  Alpco Ltd 41CO3RTE01
Encompass
The Rat Complement Factor 3 ELISA is for the quantitative determination of C3 in rat Serum or Plasma. Research Use Only. Not for Use in Diagnostic Procedures.
Catalog No. NC2885159