Learn More
Bio Rad Laboratories Rat Cytokine 23-Plex Immunoassay, 96-well kit, 23 cytokines detected, Magnetic beads, 6.5 μm
Supplier:  Bio Rad Laboratories 12005641
Encompass
The Bio-Plex Pro Rat Cytokine 23-Plex Immunoassay is a comprehensive 96-well kit for the multiplex detection of 23 rat cytokines in a single well. This immunoassay utilizes advanced technology with magnetic beads to streamline processing and enhance performance. Each kit contains premixed standards, allowing broad dynamic range generation for each target.
Catalog No. NC2541799
Only null left
Add to Cart
Request a Quote