Learn More
Raybiotech Inc Quantibody Rat Cytokine Array 1 Kit.
Supplier:  Raybiotech Inc QARCYT12
Encompass_Preferred
Detects 10 Rat Cytokines. Suitable for all liquid sample types. Quantitative, Sandwich-based, Glass Slide Antibody Array. Simultaneous quantitative measurement of multiple proteins in a wide variety of sample types. Quantibody combines the high specificity and sensitivity of ELISA with the high throughput of the glass chip-based array. Less sample, more data: only 50 ul of sample is needed for quantification of up to 40 cytokines. Proteins detected: CINC-2, CINC-3, CNTF, Fractalkine (CX3CL1), GM-CSF, IL-1 alpha (IL-1 F1), IL-4, LIX, beta-NGF, VEGF-A
Catalog No. 50-195-0904