Learn More
Sigma Aldrich Fine Chemicals Biosciences Rat Dermal fibroblasts RD
Supplier:  Sigma Aldrich Fine Chemicals Biosciences R10605N
Encompass
RDf from Cell Applications Inc. have been used in allogenic prosthetic grafts demonstrating improved epithelization collagen organization and faster regeneration (Nomoto 2012) in a cell sheet engineering study where a sheet of dermal fibroblasts was transplanted into the damaged liver in allogeneic rat model to promote migration of hepatic progenitor cells and faster healing (Muraoka 2013) and as a control in engineered cardiac tissues (Serrao 2012).
Catalog No. 50-290-4720