Learn More
American Research Products Inc Rat Dihydrotestosterone DHT E
Supplier:  American Research Products Inc CSBE07879R
Encompass
Rat Dihydrotestosterone DHT ELISA Kit from CUSABIO Detection Range 10 pg/mL-2000 pg/mL Sensitivity 5 pg/mL Sample serum plasma tissue homogenates Method Competitive
Catalog No. 50-003-26852