Learn More
American Research Products Inc Rat eosinophil cationic protei
Supplier:  American Research Products Inc CSBE11238R
Encompass
Rat eosinophil cationic protein ECP ELISA Kit from CUSABIO Detection Range 62 5 pg/mL-4000 pg/mL Sensitivity 15 6 pg/mL Sample serum plasma tissue homogenates Method Sandwich
Catalog No. 50-003-29076