Learn More
BIOHIPPO Rat GHRL Microsample
SDP
Supplier:  BIOHIPPO BHE15210762
Encompass
Rat GHRL Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of GHRL in serum plasma tissue homogenates cell lysates cell culture supernates and other biological fluids It provides a detection range of 78 13-5000 pg/mL with a sensitivity of 30 pg/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Rat GHRL Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Rat GHRL Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Rat GHRL biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color
Catalog No. 50-003-14651