Learn More
BIOHIPPO Rat HDLBP Microsample
SDP
Supplier:  BIOHIPPO BHE15210712
Encompass
Rat HDLBP Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of HDLBP in tissue homogenates cell lysates cell culture supernates and other biological fluids It provides a detection range of 0 32-20 ng/mL with a sensitivity of 0 139 ng/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Rat HDLBP Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Rat HDLBP Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Rat HDLBP biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color
Catalog No. 50-003-13914