Learn More
Electron Microscopy Sciences Rat Heart Matrix Adult, 1 mm Slices
SDP
Supplier:  Electron Microscopy Sciences 69050C
Encompass_Preferred
Constructed from high grade zinc to allow for rapid temperature equilibration, easy cleaning, and sterilization.
Size: 1 mm Slices
Catalog No. 50-283-70