Learn More
BIOHIPPO Rat hs-CRP Microsample
SDP
Supplier:  BIOHIPPO BHE15212681
Encompass
Rat hs-CRP Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of hs-CRP in serum plasma tissue homogenates cell lysates cell culture supernates and other biological fluids It provides a detection range of 1 56-100 g/mL with a sensitivity of 1 08 g/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Rat hs-CRP Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Rat hs-CRP Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Rat hs-CRP biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color
Catalog No. 50-003-15262