Learn More
Immunology Consultants Rat IgG Purified, 5.0 mg
SDP
Supplier:  Immunology Consultants AG25G
Encompass_Preferred
The IgG h+l was purified from rat serum using a combination of procedures that include salt fractionation and protein A purification. Antibody concentration was determined by extinction coefficient: O.D. 1.40 at 280nM equals 1.0 mg of IgG. Optimal working dilutions should be determined experimentally by the investigator.
Catalog No. 50-241-4307