Learn More
BIOHIPPO Rat ITGa2 Microsample
SDP
Supplier:  BIOHIPPO BHE15210958
Encompass
Rat ITGa2 Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of ITGa2 in Tissue homogenates and other biological fluids It provides a detection range of 0 32-20 ng/mL with a sensitivity of 0 112 ng/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Rat ITGa2 Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Rat ITGa2 Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Rat ITGa2 biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color
Catalog No. 50-003-15123