Learn More
BIOHIPPO Rat LEPR Microsample
SDP
Supplier:  BIOHIPPO BHE15211987
Encompass
Rat LEPR Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of LEPR in Serum plasma tissue homogenates and other biological fluids It provides a detection range of 1 57-100 ng/mL with a sensitivity of 0 64 ng/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Rat LEPR Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Rat LEPR Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Rat LEPR biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color
Catalog No. 50-003-15402