Learn More
Raybiotech Inc Rat Lipocalin-2 (NGAL) ELISA Kit
Supplier:  Raybiotech Inc ELRLIPOCALIN21
Encompass_Preferred
For cell culture supernatants, plasma, and serum samples. Sensitivity: 30 pg/mL. Pre-coated 96-well strip plate with additional reagents to allow for use in multiple experiments. Specificity: This ELISA kit shows no cross-reactivity with the following cytokines tested: rat CINC-2, CINC-3, CNTF, Fractalkine, IL-1 alpha, IL-1 beta, IL-4, IL-6, IL-10, GM-CSF, IFN-gamma, Leptin, Lix, MCP-1, MIP-3 alpha, beta-NGF, TIMP-1, TNF-alpha.
Catalog No. 50-195-0277