Learn More
BIOHIPPO Rat LOX1 Microsample
SDP
Supplier:  BIOHIPPO BHE15211248
Encompass
Rat LOX1 Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of LOX1 in Serum plasma and other biological fluids It provides a detection range of 62 5-4000 pg/mL with a sensitivity of 24 6 pg/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Rat LOX1 Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Rat LOX1 Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Rat LOX1 biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color
Catalog No. 50-003-13757