Learn More
American Research Products Inc Rat Macrophage colony stimulat
Supplier:  American Research Products Inc CSBEL006044RA
Encompass
Rat Macrophage colony-stimulating factor 1 receptor (CSF1R) ELISA kit from CUSABIO Detection Range 0 312 ng/mL-20 ng/mL Sensitivity 0 078 ng/mL Sample serum plasma tissue homogenates Method Sandwich
Catalog No. 50-003-28003