Learn More
Biomatik Corporation Rat Matrix Metalloproteinase 23B (MMP23B) ELISA Kit
Supplier:  Biomatik Corporation EKU05856
Encompass_Preferred
Rat Matrix Metalloproteinase 23B (MMP23B) ELISA Kit for detecting MMP23B in tissue homogenates, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 0.156-10ng/ml. Sensitivity: 0.071ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: O88272. Gene ID: 94339. Target Synonyms: MMP22; MMP23-B; MIFR; MIFR-1; Matrix Metalloproteinase 22; Femalysin; Matrix Metalloproteinase In The Female Reproductive Tract. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-5912