Learn More
BIOHIPPO Rat MEPE Microsample
SDP
Supplier:  BIOHIPPO BHE15210972
Encompass
Rat MEPE Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of MEPE in serum plasma tissue homogenates cell lysates cell culture supernates and other biological fluids It provides a detection range of 0 32-20 ng/mL with a sensitivity of 0 131 ng/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Rat MEPE Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Rat MEPE Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Rat MEPE biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color
Catalog No. 50-003-14973