Learn More
BIOHIPPO Rat MFAP4 Microsample
SDP
Supplier:  BIOHIPPO BHE15211139
Encompass
Rat MFAP4 Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of MFAP4 in Serum plasma tissue homogenates and other biological fluids It provides a detection range of 0 16-10 ng/mL with a sensitivity of 0 064 ng/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Rat MFAP4 Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Rat MFAP4 Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Rat MFAP4 biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color
Catalog No. 50-003-15796