Learn More
Sigma Aldrich Fine Chemicals Biosciences Neuropeptide Y (NPY) (rat, mouse) ELISA Kit | 4272.6 DA | 1 Kit
Supplier:  Sigma Aldrich Fine Chemicals Biosciences EZRMNPY27K
Encompass
Neuropeptide Y (NPY) (rat, mouse) | Purity: N/A | Mol Wt: 4272.6 | CAS No: N/A | N/A | Rat/Mouse Neuropeptide Y ELISA KitThe Neuropeptide Y (NPY) (rat, mouse) ELISA Kit quantitatively measures NPY levels in rat and mouse plasma samples, useful for neuroscience and metabolism research involving these rodent models. This ELISA utilizes a sandwich format with antibodies specific for rat/mouse NPY. Samples and standards are incubated allowing NPY to bind coated wells. After washing, detection antibody binds captured NPY. Adding enzyme substrate generates a colorimetric signal proportional to sample NPY concentration. Compared to a standard curve, unknown NPY concentrations are determined. This convenient kit requires only 20 μl sample size and provides results in 3 hours at room temperature. No sample extraction is necessary prior to running the assay.
Catalog No. NC2600920