Learn More
EUROIMMUN US INC 3P RAT/MOUSE P1NP ELISA KIT
Supplier:  EUROIMMUN US INC 3P AC33F1
Encompass_Preferred
The Immunodiagnostic Systems Limited Rat/Mouse Pinp Eia Kit Is A Competitive Enzyme Immunoassay (Eia) For The Quantitative Determination Of N-Terminal Propeptide Of Type I Procollagen (Pinp) In Rat Or Mouse Serum Or Plasma Samples. Pinp Is A Particularly Useful Marker For Monitoring The Efficacy Of Osteoporosis Therapy With Anabolic Agents, But It Is Also One Of The Best Bone Turnover Markers For Monitoring The Efficacy Of Antiresorptive Therapy. The Rat/Mouse Pinp Eia Assay Is A Specific Method To Determine Pinp Released During Rat And Mouse Bone Collagen Synthesis, And It Has No Cross-Reactivity For Human Pinp. The Rat/Mouse Pinp Eia Assay Can Be Used For Determining The Bone Formation Rate From Rat And Mouse Serum Samples1, 2, 3, And From The Culture Medium In Rat And Mouse Osteoblast Cultures.
Catalog No. NC9666468