Learn More
Raybiotech Inc C-Series Rat Neuro Discovery Array C1 Kit.
Supplier:  Raybiotech Inc AARNEU12
Encompass_Preferred
Detects 19 rat neurologically relevant proteins. Suitable for serum, plasma, cell culture media and other liquid sample types. Semi-Quantitative, Sandwich-based, Membrane Antibody Array. Screen and compare protein expression levels with high specificity and sensitivity (pg/ml levels). No dedicated equipment required. Compatible with any chemiluminescent imaging system. Proteins detected: beta-NGF, CNTF, Eotaxin-1 (CCL11), GM-CSF, IFN-gamma, IL-10, IL-1 alpha (IL-1 F1), IL-1 beta (IL-1 F2), IL-4, IL-6, LIX, MCP-1 (CCL2), Neuropilin-1, Notch-1, Notch-2, RAGE, TNF alpha, VEGF-A, BDNF
Catalog No. 50-194-8790