Learn More
BIOHIPPO Rat NPY1R Microsample
SDP
Supplier:  BIOHIPPO BHE15211046
Encompass
Rat NPY1R Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of NPY1R in Tissue homogenates and other biological fluids It provides a detection range of 0 32-20 ng/mL with a sensitivity of 0 121 ng/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Rat NPY1R Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Rat NPY1R Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Rat NPY1R biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color
Catalog No. 50-003-14977