Learn More
American Research Products Inc Rat ParaoXonase 1 PON1 ELISA
Supplier:  American Research Products Inc CSBE14340R
Encompass
Rat ParaoXonase 1 (PON1) ELISA Kit from CUSABIO Detection Range 15 6 U/mL-1000 U/mL Sensitivity 3 9 U/mL Sample serum plasma tissue homogenates Method Sandwich
Catalog No. 50-003-30201