Learn More
Bon Opus Biosciences Recombinant Rat PD-1 (C-His) 50ug
Supplier: Bon Opus Biosciences BP100150UG
Recombinant Rat PD-1 (C-His) is produced by Human Cells. The target gene encoding Leu25-Gln167 is expressed with a C-His tag. Accession# NP_001100397.1. Formulation: Lyophilized from a 0.2 μm filtered solution of PBS, pH 7.4. Bon Opus Cat# BP1001