Learn More
Discovery Life Sciences Rat (Sprague-Dawley) Pooled Liver S9 | Male | 1.0 mL
SDP
Supplier:  Discovery Life Sciences 452591
Encompass
This S9 product is prepared from sexually mature male Sprague-Dawley rat livers and characterized for key cytochrome P450 and sulfotransferase enzyme activities.
Catalog No. NC1790200