Learn More
BIOHIPPO Rat PPARa Microsample
SDP
Supplier:  BIOHIPPO BHE15211490
Encompass
Rat PPARa Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of PPARa in Tissue homogenates cell lysates cell culture supernates and other biological fluids It provides a detection range of 0 78-50 ng/mL with a sensitivity of 0 32 ng/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Rat PPARa Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Rat PPARa Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Rat PPARa biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color
Catalog No. 50-003-15873