Learn More
Braintree Scientific Flat Bottom 2.5"" dia x 6"", 125-250gm rats
SDP
Supplier:  Braintree Scientific FBML
Encompass
Economy Flat Bottomed Restrainers. Individual semi cylindrical plastic restrainers in small, medium and large. Each unit has an open slot on the bottom (for intra-peritoneal injections), a hole on the top (for the neck), and a slotted tail piece (for intravenous injections). Front slots provide ample ventilation. **Sold for research purposes only and not for human use**
Catalog No. NC9197802